Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Aminocephalosporanic acid, 98% (dry wt.), may cont. up to 2% water

$183.82 - $183.82

Chemical Identifiers

Chemical Name or Material 7-Aminocephalosporanic acid
CAS 957-68-6
Molecular Formula C10H12N2O5S
Formula Weight 272.28
MDL Number MFCD00005177
Beilstein 622638
Merck Index 14,434
Synonym 7-ACA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1053006

  SDS
alfa aesar™
A1053006
5g Each for $183.82

AAA1053003

  SDS
alfa aesar™
A1053003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1053014

  SDS
alfa aesar™
A1053014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 7-Aminocephalosporanic acid
CAS 957-68-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2°C to 8°C
Melting Point >300°C
Quantity 5g
Concentration May contain up to 2% water
Assay 98%
Assay Percent Notes (dry wt.)
Molecular Formula C10H12N2O5S
Formula Weight 272.28
MDL Number MFCD00005177
Beilstein 622638
Merck Index 14,434
Synonym 7-ACA
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

2°C to 8°C