missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 7-Amino-5-bromoindole, 97%
$179.49 - $864.08
Chemical Identifiers
CAS | 374537-99-2 |
---|---|
Molecular Formula | C8H7BrN2 |
Molecular Weight (g/mol) | 211.062 |
MDL Number | MFCD02093961 |
InChI Key | ZRTCJQGCGWZTAG-UHFFFAOYSA-N |
Synonym | 5-bromoindole-7-ylamine, 7-amino-5-bromoindole, acmc-1ae17 |
PubChem CID | 7010313 |
IUPAC Name | 5-bromo-1H-indol-7-amine |
SMILES | C1=CNC2=C(C=C(C=C21)Br)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL17795MC
|
Alfa Aesar™
L17795MC |
100mg |
Each for $179.49
|
|
AAL17795ME
|
Alfa Aesar™
L17795ME |
500mg |
Each for $528.93
|
|
AAL1779503
|
Alfa Aesar™
L1779503 |
1g |
Each for $864.08
|
|
Chemical Identifiers
374537-99-2 | |
211.062 | |
ZRTCJQGCGWZTAG-UHFFFAOYSA-N | |
7010313 | |
C1=CNC2=C(C=C(C=C21)Br)N |
C8H7BrN2 | |
MFCD02093961 | |
5-bromoindole-7-ylamine, 7-amino-5-bromoindole, acmc-1ae17 | |
5-bromo-1H-indol-7-amine |
Specifications
Air sensitive | |
100mg | |
C8H7BrN2 | |
UN2811 | |
ZRTCJQGCGWZTAG-UHFFFAOYSA-N | |
5-bromo-1H-indol-7-amine | |
7010313 | |
97% |
104°C to 105°C | |
374537-99-2 | |
MFCD02093961 | |
5-bromoindole-7-ylamine, 7-amino-5-bromoindole, acmc-1ae17 | |
C1=CNC2=C(C=C(C=C21)Br)N | |
211.062 | |
211.06 | |
7-Amino-5-bromoindole |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Store at -20°C