missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Amino-4-methylcoumarin [for HPLC Labeling], TCI America™

Manufacturer:  TCI America M07601G

 View more versions of this product

Catalog No. M07601G



Specifications

Specifications

7-Amino-4-methylcoumarin [for HPLC Labeling]
228°C
26093-31-2
MFCD00006868
GLNDAGDHSLMOKX-UHFFFAOYSA-N
7-amino-4-methylchromen-2-one
92249
175.19
≥98.0% (HPLC,T)
Undesignated
1g
C10H9NO2
7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
CC1=CC(=O)OC2=C1C=CC(=C2)N
175.187
CHEBI:51771
Crystal-Powder at 20°C

Chemical Identifiers

26093-31-2
175.187
GLNDAGDHSLMOKX-UHFFFAOYSA-N
92249
7-amino-4-methylchromen-2-one
C10H9NO2
MFCD00006868
7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
CHEBI:51771
CC1=CC(=O)OC2=C1C=CC(=C2)N
Safety and Handling

Safety and Handling

TSCA : Yes

Documents