Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Amino-4-(methoxymethyl)coumarin, 97%

$32.09 - $89.04

Chemical Identifiers

Chemical Name or Material 7-Amino-4-(methoxymethyl)coumarin
CAS 175205-10-4
Molecular Formula C11H11NO3
Formula Weight 205.22
MDL Number MFCD00068088
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1853201

  SDS
alfa aesar™
A1853201
0.25g Each for $32.09

AAA1853203

  SDS
alfa aesar™
A1853203
1g Each for $89.04
Description & Specifications

Specifications

Formula Weight 205.22
Assay 97%
CAS 175205-10-4
Melting Point 181° to 186°C
Quantity 0.25g
Chemical Name or Material 7-Amino-4-(methoxymethyl)coumarin
MDL Number MFCD00068088
Molecular Formula C11H11NO3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.