missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Amino-1H-indazole, 95%, ACROS Organics™

Chemical Identifiers

CAS 21443-96-9
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00022790
InChI Key OTFFCAGPSWJBDK-UHFFFAOYSA-N
Synonym 1h-indazol-7-amine, 1, 1h-indazol-7-ylamine, 1h-indazole-7-ylamine, 2h-indazol-7-amine, 7-aminindazole, 7-amino-1h-indazole, 7-aminoindazole, cbi-bb zero/005543, pubchem20891
PubChem CID 88901
IUPAC Name 1H-indazol-7-amine
SMILES C1=CC2=C(C(=C1)N)NN=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC429390010
SDS View Documents
Thermo Fisher Scientific
AC429390010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429390050
SDS View Documents
Thermo Fisher Scientific
AC429390050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

21443-96-9
133.15
OTFFCAGPSWJBDK-UHFFFAOYSA-N
88901
C1=CC2=C(C(=C1)N)NN=C2
C7H7N3
MFCD00022790
1h-indazol-7-amine, 1, 1h-indazol-7-ylamine, 1h-indazole-7-ylamine, 2h-indazol-7-amine, 7-aminindazole, 7-amino-1h-indazole, 7-aminoindazole, cbi-bb zero/005543, pubchem20891
1H-indazol-7-amine

Specifications

Authentic
Tan
1g
94.0
94% min. (HPLC)
MFCD00022790
OTFFCAGPSWJBDK-UHFFFAOYSA-N
1H-indazol-7-amine
88901
Crystalline Solid
Glass Bottle
164.0°C to 164.5°C
21443-96-9
100.0
C7H7N3
1h-indazol-7-amine, 1, 1h-indazol-7-ylamine, 1h-indazole-7-ylamine, 2h-indazol-7-amine, 7-aminindazole, 7-amino-1h-indazole, 7-aminoindazole, cbi-bb zero/005543, pubchem20891
C1=CC2=C(C(=C1)N)NN=C2
133.15
133.15
95%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-391-8