Please login to your online account to display your discounted pricing

7-Amino-1-methyl-1H-indazole, 95%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC432440010

 View more versions of this product

Catalog No. AC432440010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥94.0%
Formula Weight 147.18
Quantity 1g
Color Yellow to brown
Melting Point 133° to 137°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 7-Amino-1-methyl-1H-indazole
Molecular Formula C8H9N3
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
MDL Number MFCD09870043
CAS 41926-06-1
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 7-Amino-1-methyl-1H-indazole
CAS 41926-06-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C8H9N3
Formula Weight 147.18
MDL Number MFCD09870043
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents