missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Amino-1-methyl-1H-indazole, 95%, ACROS Organics™

Chemical Identifiers

CAS 41926-06-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H9N3
Formula Weight 147.18
MDL Number MFCD09870043
Chemical Name or Material 7-Amino-1-methyl-1H-indazole
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC432440010 SDS Thermo Fisher Scientific
AC432440010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC432440050 SDS Thermo Fisher Scientific
AC432440050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

41926-06-1
100.0
C8H9N3
MFCD09870043
≥94.0%
94.0
95%
147.18
7-Amino-1-methyl-1H-indazole

Specifications

41926-06-1
94.0
133° to 137°C
Crystalline Powder
95%
147.18
7-Amino-1-methyl-1H-indazole
Authentic
100.0
Yellow to brown
1g
C8H9N3
MFCD09870043
≥94.0%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.