missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Amino-1,3-naphthalenedisulfonic acid, Tech., ACROS Organics™

Manufacturer:  Acros Organics 163965000

 View more versions of this product

Catalog No. AC163965000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

7-Amino-1, 3-naphthalenedisulfonic acid
Glass bottle
500g
86-65-7
MFCD00003991
15, 395
CMOLPZZVECHXKN-UHFFFAOYSA-N
7-aminonaphthalene-1,3-disulfonic acid
6851
Powder or Needles
Technical
Authentic
Brown to White
tech.
C10H9NO6S2
14, 784
amido-g-acid, 7-amino-1,3-naphthalenedisulfonic acid, amino-g-acid, 1,3-naphthalenedisulfonic acid, 7-amino, amino-g acid, 2-amino-6,8-disulfonaphthalene, unii-7c482fx29k, 2-aminonaphthalene-6,8-disulfonic acid, 2-naphthylamine-6,8-disulfonic acid, beta-naphthylamine-6,8-disulfonic acid
C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N
303.303
303.29
60% (as is basis) Typically

Chemical Identifiers

86-65-7
303.303
CMOLPZZVECHXKN-UHFFFAOYSA-N
6851
C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N
C10H9NO6S2
MFCD00003991
amido-g-acid, 7-amino-1,3-naphthalenedisulfonic acid, amino-g-acid, 1,3-naphthalenedisulfonic acid, 7-amino, amino-g acid, 2-amino-6,8-disulfonaphthalene, unii-7c482fx29k, 2-aminonaphthalene-6,8-disulfonic acid, 2-naphthylamine-6,8-disulfonic acid, beta-naphthylamine-6,8-disulfonic acid
7-aminonaphthalene-1,3-disulfonic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-689-2

RTECSNumber : QJ6132000

TSCA : TSCA

SDS Documents