missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Acetyl-8-bromo-3,4-dihydro-2H-1,5-benzodioxepine

$91.91 - $91.91

Chemical Identifiers

CAS 175136-35-3
Molecular Formula C11H11BrO3
Formula Weight 271.11
MDL Number MFCD00218807
Chemical Name or Material 7-Acetyl-8-bromo-3,4-dihydro-2H-1,5-benzodioxepine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50591MD SDS Alfa Aesar™
H50591MD
250mg Each for $91.91
AAH5059103 SDS Alfa Aesar™
H5059103
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

175136-35-3
271.11
7-Acetyl-8-bromo-3,4-dihydro-2H-1,5-benzodioxepine
C11H11BrO3
MFCD00218807

Specifications

175136-35-3
271.11
250mg
C11H11BrO3
MFCD00218807
7-Acetyl-8-bromo-3,4-dihydro-2H-1,5-benzodioxepine