missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Acetyl-3,4-dihydro-1,5-benzodioxepin, 96%, Thermo Scientific™

$49.68 - $229.67

Chemical Identifiers

CAS 22776-09-6
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00114700
InChI Key VGRSZNPWUZDPCH-UHFFFAOYSA-N
Synonym 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethan-1-one, 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4-dioxepin-7-yl ethanone, 3,4-trimethylenedioxyacetophenone, 7-acetyl-2h,3h,4h-benzo b 1,4-dioxepin, 7-acetyl-3,4-dihydro-1,5-benzodioxepin, 7-acetyl-3,4-dihydro-2h-1,5-benzodioxepin, acmc-20apc2, zlchem 1110
PubChem CID 2775272
IUPAC Name 1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)OCCCO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2704003
View Documents Promotion Details
Thermo Fisher Scientific
H2704003
1g
Each for $49.68
 
AAH2704009
View Documents Promotion Details
Thermo Fisher Scientific
H2704009
10g
Each for $229.67
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

22776-09-6
192.214
VGRSZNPWUZDPCH-UHFFFAOYSA-N
2775272
CC(=O)C1=CC2=C(C=C1)OCCCO2
C11H12O3
MFCD00114700
1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethan-1-one, 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4-dioxepin-7-yl ethanone, 3,4-trimethylenedioxyacetophenone, 7-acetyl-2h,3h,4h-benzo b 1,4-dioxepin, 7-acetyl-3,4-dihydro-1,5-benzodioxepin, 7-acetyl-3,4-dihydro-2h-1,5-benzodioxepin, acmc-20apc2, zlchem 1110
1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethanone

Specifications

22776-09-6
C11H12O3
1g
VGRSZNPWUZDPCH-UHFFFAOYSA-N
1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethanone
2775272
96%
120°C (0.5mmHg)
MFCD00114700
1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethan-1-one, 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4-dioxepin-7-yl ethanone, 3,4-trimethylenedioxyacetophenone, 7-acetyl-2h,3h,4h-benzo b 1,4-dioxepin, 7-acetyl-3,4-dihydro-1,5-benzodioxepin, 7-acetyl-3,4-dihydro-2h-1,5-benzodioxepin, acmc-20apc2, zlchem 1110
CC(=O)C1=CC2=C(C=C1)OCCCO2
192.214
192.21
7-Acetyl-3,4-dihydro-1,5-benzodioxepin
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications