Please login to your online account to display your discounted pricing

7,8-Dihydroxyflavone, Tocris™

TrkB agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material 7, 8-Dihydroxyflavone
Assay Percent Range >98%
Molecular Formula C15H10O4
Formula Weight 254.24
Synonym 7, 8-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

382650

 
r & d systems
RDS382650
50mg Each for $862.95

382610

 
r & d systems
RDS382610
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 254.24
Quantity 50mg
Chemical Name or Material 7, 8-Dihydroxyflavone
Assay Percent Range >98%
Molecular Formula C15H10O4
CAS 38183-03-8
Synonym 7, 8-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C