Please login to your online account to display your discounted pricing

Alfa Aesar™ 7,8-Dichloro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Chemical Identifiers

Chemical Name or Material 7,8-Dichloro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS 300675-28-9
Formula Weight 258.06
MDL Number MFCD01911608
Synonym CAY10577
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65324LB0

 
alfa aesar™
J65324LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.06
Chemical Name or Material 7,8-Dichloro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonym CAY10577
MDL Number MFCD01911608
Physical Form Crystalline Solid
Storage Note 1 -30° to -10°C
Quantity 5mg
CAS 300675-28-9
Safety and Handling

-30° to -10°C