Please login to your online account to display your discounted pricing

Alfa Aesar™ 7,7-Dimethyleicosadienoic acid, 99%


Quantity 10mg

Manufacturer: alfa aesar™  J67249MA

 View more versions of this product

Catalog No. AAJ67249MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 336.56
Chemical Name or Material 7,7-Dimethyleicosadienoic acid
MDL Number MFCD00133010
Assay 99%
CAS 89560-01-0
Synonym DEDA
Quantity 10mg
Color Colorless to pale yellow
Physical Form Viscous Oil
Storage Note 1 2° to 8°C

Specifications

Chemical Name or Material 7,7-Dimethyleicosadienoic acid
CAS 89560-01-0
Formula Weight 336.56
MDL Number MFCD00133010
Synonym DEDA
Accessories

2° to 8°C