Please login to your online account to display your discounted pricing

Alfa Aesar™ 7,7-Dimethyleicosadienoic acid, 99%

Chemical Identifiers

Chemical Name or Material 7,7-Dimethyleicosadienoic acid
CAS 89560-01-0
Formula Weight 336.56
MDL Number MFCD00133010
Synonym DEDA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67249MA

 
alfa aesar™
J67249MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 336.56
Chemical Name or Material 7,7-Dimethyleicosadienoic acid
MDL Number MFCD00133010
CAS 89560-01-0
Assay 99%
Synonym DEDA
Storage Note 1 2° to 8°C
Physical Form Viscous Oil
Quantity 10mg
Color Colorless to pale yellow
Safety and Handling

2° to 8°C