Please login to your online account to display your discounted pricing

Alfa Aesar™ 7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one, 97%

$267.84 - $267.84

Chemical Identifiers

Chemical Name or Material 7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one
CAS 5307-99-3
Molecular Formula C7H6Cl2O
Formula Weight 177.03
MDL Number MFCD00013738
Beilstein 2045059
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1791514

  SDS
alfa aesar™
A1791514
25g Each for $267.84
Description & Specifications

Specifications

Formula Weight 177.03
Chemical Name or Material 7,7-Dichlorobicyclo[3.2.0]hept-2-en-6-one
Assay 97%
Molecular Formula C7H6Cl2O
MDL Number MFCD00013738
CAS 5307-99-3
Boiling Point 127° to 128°C (25mmHg)
Beilstein 2045059
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Recommended Storage 2° to 8°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C