missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7,7,8,8-Tetracyanoquinodimethane, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC169360500

 View more versions of this product

Catalog No. AC169360500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

7,7,8,8-Tetracyanoquinodimethane
Glass bottle
50g
1518-16-7
100.0
C12H4N4
01,1136; 12,464
PCCVSPMFGIFTHU-UHFFFAOYSA-N
2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
73697
204.19
≥97.5%
Authentic
Brown-Orange to Green
98%
97.5
98%
MFCD00011664
7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N
204.192
CHEBI:52445
Crystalline Powder

Chemical Identifiers

1518-16-7
204.192
PCCVSPMFGIFTHU-UHFFFAOYSA-N
73697
2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
C12H4N4
MFCD00011664
7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
CHEBI:52445
C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-174-8

RTECSNumber : GU4850000

TSCA : TSCA

SDS Documents