Please login to your online account to display your discounted pricing

7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC78239DA

 View more versions of this product

Catalog No. CC78239DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 244.1
Quantity 1g
Assay 97%
CAS 1192755-14-8
Packaging Amber Glass Bottle
Chemical Name or Material 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Molecular Formula C14H17BO3

Specifications

Chemical Name or Material 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Assay 97%
Molecular Formula C14H17BO3
Formula Weight 244.1