Please login to your online account to display your discounted pricing

7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan, Maybridge

Chemical Identifiers

Chemical Name or Material 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Assay 97%
Molecular Formula C14H17BO3
Formula Weight 244.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC78239DE

 
fisher scientific company
CC78239DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC78239DA

 
fisher scientific company
CC78239DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.1
Quantity 5g
Assay 97%
Molecular Formula C14H17BO3
Packaging Amber Glass Bottle
CAS 1192755-14-8
Chemical Name or Material 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan