Please login to your online account to display your discounted pricing

7,12-Dimethylbenz[a]anthracene, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC408180050

 View more versions of this product

Catalog No. AC408180050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 256.34
Color Yellow to greenish
Quantity 5g
Beilstein 05, III, 2413
Merck Index 15, 3257
EINECS Number 200-359-5
TSCA TSCA
RTECS Number CW3850000
Melting Point 121° to 124°C
CAS Max % 100.0
Synonym 1, 4-Dimethyl-2, 3-benzphenanthrene
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause cancer.
Chemical Name or Material 7, 12-Dimethylbenz[a]anthracene
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C20H16
MDL Number MFCD00003600
CAS 57-97-6
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 7, 12-Dimethylbenz[a]anthracene
Name Note 99%
CAS 57-97-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C20H16
Formula Weight 256.34
MDL Number MFCD00003600
Synonym 1, 4-Dimethyl-2, 3-benzphenanthrene
Accessories

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause cancer.

SDS & Safety Documents