missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 83-43-2 |
---|---|
Molecular Formula | C22H30O5 |
Molecular Weight (g/mol) | 374.477 |
MDL Number | MFCD00010591 |
InChI Key | VHRSUDSXCMQTMA-PJHHCJLFSA-N |
Synonym | methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol |
PubChem CID | 6741 |
ChEBI | CHEBI:6888 |
IUPAC Name | (6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one |
SMILES | CC1CC2C3CCC(C3(CC(C2C4(C1=CC(=O)C=C4)C)O)C)(C(=O)CO)O |
Chemical Identifiers
83-43-2 | |
374.477 | |
VHRSUDSXCMQTMA-PJHHCJLFSA-N | |
6741 | |
(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one |
C22H30O5 | |
MFCD00010591 | |
methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol | |
CHEBI:6888 | |
CC1CC2C3CCC(C3(CC(C2C4(C1=CC(=O)C=C4)C)O)C)(C(=O)CO)O |
Specifications
White | |
1g | |
C22H30O5 | |
methylprednisolone, medrol, medrone, urbason, methylprednisolon, 6alpha-methylprednisolone, metilbetasone, medrate, metilprednisolone, dopomedrol | |
CC1CC2C3CCC(C3(CC(C2C4(C1=CC(=O)C=C4)C)O)C)(C(=O)CO)O | |
374.477 | |
CHEBI:6888 | |
Crystal-Powder at 20°C | |
6alpha-Methylprednisolone |
237°C | |
83-43-2 | |
MFCD00010591 | |
VHRSUDSXCMQTMA-PJHHCJLFSA-N | |
(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one | |
6741 | |
374.48 | |
≥98.0% (HPLC) |
Safety and Handling
RTECSNumber : TU4146000
TSCA : Yes