Please login to your online account to display your discounted pricing

680C91, Tocris™

Potent and selective inhibitor of tryptophan 2,3-dioxygenase

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material 680C91
Assay Percent Range >98%
Molecular Formula C15H11FN2
Formula Weight 238.26
Synonym 6-Fluoro-3-[(1E)-2-(3-pyridinyl)ethenyl]-1H-indole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

439210

 
r & d systems
RDS439210
10mg Each for $221.01

439250

 
r & d systems
RDS439250
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 238.26
Quantity 10mg
Synonym 6-Fluoro-3-[(1E)-2-(3-pyridinyl)ethenyl]-1H-indole
CAS 163239-22-3
Recommended Storage Store at +4°C
Chemical Name or Material 680C91
Molecular Formula C15H11FN2
Assay Percent Range >98%
Safety and Handling

Store at +4°C