Please login to your online account to display your discounted pricing

Whatman™ 602H 1/2 Qualitative Folded Filter Paper Circle

Dense filter paper forr collecting and removing fine precipitates

$78.04 - $382.20

Specifications

Format Circle
Grade 602H 1/2
Material Cellulose
For Use With (Application) Collecting very small particles and removing fine precipitates, used in sample preparation (e.g., in the beverage industry for residual sugar determination, Acidic spectra, refractometric analysis and HPLC)
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Price Quantity    

09-924-306

 
whatman™
WHT10312642
90mm Pack of 100 Each for $78.04

09-924-307

 
whatman™
WHT10312644
125mm Pack of 100 Each for $105.07

09-924-308

 
whatman™
WHT10312645*
150mm Pack of 100 Each for $154.84

09-924-310

 
whatman™
WHT10312651
240mm Pack of 100 Each for $382.20

09-924-309

 
whatman™
WHT10312647
185mm Pack of 100 Each for $234.46
Description & Specifications

Specifications

Format Circle
Grade 602H 1/2
Material Cellulose
For Use With (Application) Collecting very small particles and removing fine precipitates, used in sample preparation (e.g., in the beverage industry for residual sugar determination, Acidic spectra, refractometric analysis and HPLC)

  • Used in sample preparation
  • Residual sugar determination
  • Acidic spectra
  • Refractometric analysis
  • Slow flow speed
  • HPLC