Promega 6 x 5 LC-MS/MS Peptide Reference Mix DFS Item
The 6 x 5 LC-MS/MS Peptide Reference Mix is a unique reagent designed to monitor liquid chromatography (LC) and mass spectrometry (MS) instrument performance and assist in method development and optimization.
Supplier:  Promega V7495
 View more versions of this product
Catalog No. PRV7495
Description
Description
  • Peptide Mix and Free Software to Monitor LC/MS Instrument Performance
  • Determine mass spec instrument sensitivity and dynamic range
  • No manual calculations, maximized instrument performance with free PReMiS™ Software
Specifications
Specifications
-30°C to -10°C
200 pmol