missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(Trifluoromethyl)uracil, 97%

Manufacturer:  Alfa Aesar™ 4446703

 View more versions of this product

Catalog No. AA4446703



Specifications

Specifications

6- (Trifluoromethyl)uracil
672-45-7
MFCD01011762
IROWWTVZNHKLLE-UHFFFAOYSA-N
6-(trifluoromethyl)-1H-pyrimidine-2,4-dione
233890
Powder
1g
C5H3F3N2O2
2,4 1h,3h-pyrimidinedione, 6-trifluoromethyl
C1=C(NC(=O)NC1=O)C(F)(F)F
180.086
180.08
97%

Chemical Identifiers

672-45-7
180.086
IROWWTVZNHKLLE-UHFFFAOYSA-N
233890
C1=C(NC(=O)NC1=O)C(F)(F)F
C5H3F3N2O2
MFCD01011762
2,4 1h,3h-pyrimidinedione, 6-trifluoromethyl
6-(trifluoromethyl)-1H-pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents