Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-(Trifluoromethyl)nicotinoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 6-(Trifluoromethyl)nicotinoyl chloride
CAS 358780-13-9
Molecular Formula C7H3ClF3NO
Formula Weight 209.56g/mol
MDL Number MFCD01862645
UN Number UN3265
Synonym 6-(Trifluoromethyl)pyridine-3-carbonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2699603

 
alfa aesar™
H26996-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2699606

 
alfa aesar™
H26996-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Boiling Point 52° to 54°C (2mmHg)
Quantity 1g
Flash Point 109°C (228°F)
Chemical Name or Material 6-(Trifluoromethyl)nicotinoyl chloride
CAS 358780-13-9
Assay 97%
Molecular Formula C7H3ClF3NO
Formula Weight 209.56g/mol
MDL Number MFCD01862645
UN Number UN3265
Synonym 6-(Trifluoromethyl)pyridine-3-carbonyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C