missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Trifluoromethyl-4-pyrimidinol, TCI America™

Manufacturer:  TCI America T160125G

 View more versions of this product

Catalog No. T160125G



Specifications

Specifications

6-Trifluoromethyl-4-pyrimidinol
170°C
1546-78-7
MFCD00192530
LVOYSBZJJWPUBD-UHFFFAOYSA-N
6-(trifluoromethyl)-1H-pyrimidin-4-one
1268071
Crystal-Powder at 20°C
White-Yellow
25g
C5H3F3N2O
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl
C1=C(NC=NC1=O)C(F)(F)F
164.087
164.09
≥98.0% (GC,T)

Chemical Identifiers

1546-78-7
164.087
LVOYSBZJJWPUBD-UHFFFAOYSA-N
1268071
C1=C(NC=NC1=O)C(F)(F)F
C5H3F3N2O
MFCD00192530
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl
6-(trifluoromethyl)-1H-pyrimidin-4-one
Safety and Handling

Safety and Handling

TSCA : No

Documents