Please login to your online account to display your discounted pricing

6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde
Assay 95%
Molecular Formula C11H13NO3
Formula Weight 207.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC58204DE

  SDS
fisher scientific company
CC58204DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC58204DA

  SDS
fisher scientific company
CC58204DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC58204CB

  SDS
fisher scientific company
CC58204CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.23
Quantity 5g
Assay 95%
Molecular Formula C11H13NO3
CAS 898289-54-8
Chemical Name or Material 6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning