missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-tert-butyl-m-cresol, 98%, ACROS Organics™

$199.64 - $199.64

Chemical Identifiers

CAS 88-60-8
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00002308
Synonym 2-tert-Butyl-5-methylphenol
Chemical Name or Material 6-tert-Butyl-m-cresol
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC403515000 SDS Thermo Fisher Scientific
AC403515000
500g Each for $199.64
Specifications Description & Specifications

Chemical Identifiers

88-60-8
100.0
C11H16O
MFCD00002308
6-tert-Butyl-m-cresol
97.5
98%
164.25
2-tert-Butyl-5-methylphenol
≥97.5%

Specifications

88-60-8
100.0
Authentic
Clear colorless
500g
C11H16O
MFCD00002308
6-tert-Butyl-m-cresol
97.5
0.9640g/mL
1.5170 to 1.5190 (20°C, 589nm)
Viscous Liquid
98%
164.25
2-tert-Butyl-5-methylphenol
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger