missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444660050
Specifications
6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester | |
5g | |
96% min. (HPLC) | |
2-boc-amino pyridine-5-boronic acid pinacol ester, 2-boc-amino pyridine-5-boronic acid, pinacol ester, 2-tert-butoxycarbonylamino pyridine-5-boronic acid, pinacol ester, boc-6-aminopyridine-3-boronic acid pinacol ester, carbamic acid,n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-,1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NC(=O)OC(C)(C)C | |
320.2 | |
320.2 |
203.0°C to 205.0°C | |
910462-31-6 | |
C16H25BN2O4 | |
USZVCDRKIVKICD-UHFFFAOYSA-N | |
tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate | |
17750222 | |
97% |
Chemical Identifiers
910462-31-6 | |
320.2 | |
2-boc-amino pyridine-5-boronic acid pinacol ester, 2-boc-amino pyridine-5-boronic acid, pinacol ester, 2-tert-butoxycarbonylamino pyridine-5-boronic acid, pinacol ester, boc-6-aminopyridine-3-boronic acid pinacol ester, carbamic acid,n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-,1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate | |
tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate |
C16H25BN2O4 | |
USZVCDRKIVKICD-UHFFFAOYSA-N | |
17750222 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NC(=O)OC(C)(C)C |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning