missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 444660010

 View more versions of this product

Catalog No. AC444660010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester
96% min. (HPLC)
C16H25BN2O4
tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-tert-butoxycarbonylamino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, boc-6-aminopyridine-3-boronic acid pinacol ester, carbamic acid,n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-,1,1-dimethylethyl ester, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NC(=O)OC(C)(C)C
320.2
320.2
910462-31-6
Glass Bottle
1g
USZVCDRKIVKICD-UHFFFAOYSA-N
tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
17750222
97%

Chemical Identifiers

910462-31-6
320.2
tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-tert-butoxycarbonylamino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, boc-6-aminopyridine-3-boronic acid pinacol ester, carbamic acid,n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-,1,1-dimethylethyl ester, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate
tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
C16H25BN2O4
USZVCDRKIVKICD-UHFFFAOYSA-N
17750222
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NC(=O)OC(C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only