missing translation for 'onlineSavingsMsg'
Learn More

[6]-Shogaol, ≥90.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014599
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014599 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014599 Supplier Merck Emd Millipore Supplier No. 3930310MG
Only null left

[6]-Shogaol is a pungent and one of the most abundant dehydrated form of gingerols present in the fresh ginger roots or dried and thermally treated roots. It may serve as a potential anti-inflammatory agent.

Featured Industries: Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 555-66-8
Physical Form Neat
Linear Formula C17H24O3
MDL Number MFCD01736094
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-4-decen-3-one
Type Analytical Standard
Quantity 10 mg
Formula Weight 276.37
Percent Purity ≥90.0% (HPLC)
Grade Analytical Standard
Show More Show Less