missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Quinoxalinol, TCI America™

Manufacturer:  TCI America Q01011G

 View more versions of this product

Catalog No. Q01011G



Specifications

Specifications

6-Quinoxalinol
252°C
7467-91-6
MFCD07364440
VEGOESHHZXUHMI-UHFFFAOYSA-N
4H-quinoxalin-6-one
343966
Crystal-Powder at 20°C
Undesignated
1g
C8H6N2O
6-Hydroxyquinoxaline
C1=CC2=NC=CNC2=CC1=O
146.149
146.15
≥98.0% (GC,T)

Chemical Identifiers

7467-91-6
146.149
VEGOESHHZXUHMI-UHFFFAOYSA-N
343966
C1=CC2=NC=CNC2=CC1=O
C8H6N2O
MFCD07364440
6-Hydroxyquinoxaline
4H-quinoxalin-6-one
Safety and Handling

Safety and Handling

TSCA : No

Documents