missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Quinoxalinol 98.0+%, TCI America™
$184.14 - $733.29
Chemical Identifiers
CAS | 7467-91-6 |
---|---|
Molecular Formula | C8H6N2O |
Molecular Weight (g/mol) | 146.15 |
MDL Number | MFCD07364440 |
InChI Key | VEGOESHHZXUHMI-UHFFFAOYSA-N |
Synonym | 6-Hydroxyquinoxaline |
PubChem CID | 343966 |
IUPAC Name | 4,6-dihydroquinoxalin-6-one |
SMILES | O=C1C=CC2=NC=CNC2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
Q0101200MG
|
TCI America
Q0101200MG |
200 mg |
Each for $184.14
|
|
|||||
Q01011G
|
TCI America
Q01011G |
1 g |
Each for $733.29
|
|
|||||
Chemical Identifiers
7467-91-6 | |
146.15 | |
VEGOESHHZXUHMI-UHFFFAOYSA-N | |
343966 | |
O=C1C=CC2=NC=CNC2=C1 |
C8H6N2O | |
MFCD07364440 | |
6-Hydroxyquinoxaline | |
4,6-dihydroquinoxalin-6-one |
Specifications
7467-91-6 | |
C8H6N2O | |
200 mg | |
VEGOESHHZXUHMI-UHFFFAOYSA-N | |
4,6-dihydroquinoxalin-6-one | |
343966 | |
≥98.0% (GC,T) | |
6-Quinoxalinol |
252°C | |
MFCD07364440 | |
6-Hydroxyquinoxaline | |
O=C1C=CC2=NC=CNC2=C1 | |
146.15 | |
146.15 | |
Crystalline Powder |
Safety and Handling
TSCA : No