Please login to your online account to display your discounted pricing

6-Quinoxalinecarbonyl chloride, 95%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC11702CB

 View more versions of this product

Catalog No. CC11702CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 192.6
Quantity 250mg
Assay 97%
CAS Max % 100.0
Packaging Amber glass bottle
Health Hazard 1 GHS Signal Word: Danger
CAS 258503-93-4
Chemical Name or Material 6-Quinoxalinecarbonyl chloride
Assay Percent Range 95%
CAS Min % 96.5
Molecular Formula C9H5ClN2O
MDL Number MFCD02682002
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 6-Quinoxalinecarbonyl chloride
CAS 258503-93-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 95%
Molecular Formula C9H5ClN2O
Formula Weight 192.6
MDL Number MFCD02682002
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents