missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Quinoxalinecarbonyl chloride, 95%, Maybridge™

$85.61 - $85.61

Chemical Identifiers

CAS 258503-93-4
Molecular Formula C9H5ClN2O
Molecular Weight (g/mol) 192.60
MDL Number MFCD02682002
InChI Key UTEQROVYZDJSOO-UHFFFAOYSA-N
Synonym 6-chlorocarbonyl quinoxaline, 6-quinoxaline carbonyl chloride, 6-quinoxalinecarbonyl chloride, 6-quinoxalinecarbonyl chloride 9ci, 6-quinoxalinecarbonylchloride
PubChem CID 2776328
IUPAC Name quinoxaline-6-carbonyl chloride
SMILES ClC(=O)C1=CC=C2N=CC=NC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC11702CB
SDS View Documents Promotion Details
Fisher Scientific Company
CC11702CB
250mg
Each for $85.61
 
Description

Description

Specifications

Chemical Identifiers

258503-93-4
192.60
UTEQROVYZDJSOO-UHFFFAOYSA-N
2776328
ClC(=O)C1=CC=C2N=CC=NC2=C1
C9H5ClN2O
MFCD02682002
6-chlorocarbonyl quinoxaline, 6-quinoxaline carbonyl chloride, 6-quinoxalinecarbonyl chloride, 6-quinoxalinecarbonyl chloride 9ci, 6-quinoxalinecarbonylchloride
quinoxaline-6-carbonyl chloride

Specifications

Amber glass bottle
258503-93-4
100.0
C9H5ClN2O
6-chlorocarbonyl quinoxaline, 6-quinoxaline carbonyl chloride, 6-quinoxalinecarbonyl chloride, 6-quinoxalinecarbonyl chloride 9ci, 6-quinoxalinecarbonylchloride
ClC(=O)C1=CC=C2N=CC=NC2=C1
192.60
192.6
6-Quinoxalinecarbonyl chloride
250mg
96.5
95%
MFCD02682002
UTEQROVYZDJSOO-UHFFFAOYSA-N
quinoxaline-6-carbonyl chloride
2776328
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications