missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Quinoxalinecarbonyl chloride, 95%, Maybridge

Manufacturer:  Fisher Scientific Company CC11702DE

 View more versions of this product

Catalog No. CC11702DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

192.6
97%
6-Quinoxalinecarbonyl chloride
96.5
C9H5ClN2O
5g
100.0
95%
MFCD02682002
258503-93-4

Chemical Identifiers

258503-93-4
100.0
C9H5ClN2O
MFCD02682002
97%
96.5
95%
192.6
6-Quinoxalinecarbonyl chloride

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger