missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Quinoxalinamine, 97%, Maybridge

Chemical Identifiers

CAS 6298-37-9
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.165
MDL Number MFCD00462821
InChI Key MSGRFBKVMUKEGZ-UHFFFAOYSA-N
Synonym 6-amino-quinoxaline
PubChem CID 237859
IUPAC Name quinoxalin-6-amine
SMILES C1=CC2=NC=CN=C2C=C1N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC11714EA SDS View Documents Fisher Scientific Company
CC11714EA
10g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

6298-37-9
145.165
MSGRFBKVMUKEGZ-UHFFFAOYSA-N
237859
C1=CC2=NC=CN=C2C=C1N
C8H7N3
MFCD00462821
6-amino-quinoxaline
quinoxalin-6-amine

Specifications

Amber Glass Bottle
6298-37-9
100.0
C8H7N3
6-amino-quinoxaline
C1=CC2=NC=CN=C2C=C1N
145.165
145.16
6-Quinoxalinamine
10g
96.5
97%
MFCD00462821
MSGRFBKVMUKEGZ-UHFFFAOYSA-N
quinoxalin-6-amine
237859
97%