missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Pyrrolidin-1-ylpyridine-2-carbaldehyde, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 230618-24-3 |
---|---|
Molecular Formula | C10H12N2O |
Molecular Weight (g/mol) | 176.22 |
MDL Number | MFCD08435859 |
InChI Key | SZOZYXCTOMKVTL-UHFFFAOYSA-N |
Synonym | 6-pyrrolidin-1-yl picolinaldehyde, 6-pyrrolidin-1-yl pyridine-2-carbaldehyde, 6-pyrrolidin-1-yl-picolinaldehyde, 6-pyrrolidinylpyridine-2-carbaldehyde, 6-1-pyrrolidinyl-2-pyridinecarboxaldehyde, 2-pyridinecarboxaldehyde,6-1-pyrrolidinyl, 2-pyridinecarboxaldehyde, 6-1-pyrrolidinyl |
PubChem CID | 10821162 |
IUPAC Name | 6-pyrrolidin-1-ylpyridine-2-carbaldehyde |
SMILES | O=CC1=NC(=CC=C1)N1CCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC38004CB
|
Thermo Scientific Chemicals
CC38004CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC38004DA
|
Thermo Scientific Chemicals
CC38004DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC38004EA
|
Thermo Scientific Chemicals
CC38004EA |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
230618-24-3 | |
176.22 | |
SZOZYXCTOMKVTL-UHFFFAOYSA-N | |
10821162 | |
O=CC1=NC(=CC=C1)N1CCCC1 |
C10H12N2O | |
MFCD08435859 | |
6-pyrrolidin-1-yl picolinaldehyde, 6-pyrrolidin-1-yl pyridine-2-carbaldehyde, 6-pyrrolidin-1-yl-picolinaldehyde, 6-pyrrolidinylpyridine-2-carbaldehyde, 6-1-pyrrolidinyl-2-pyridinecarboxaldehyde, 2-pyridinecarboxaldehyde,6-1-pyrrolidinyl, 2-pyridinecarboxaldehyde, 6-1-pyrrolidinyl | |
6-pyrrolidin-1-ylpyridine-2-carbaldehyde |
Specifications
230618-24-3 | |
C10H12N2O | |
250mg | |
SZOZYXCTOMKVTL-UHFFFAOYSA-N | |
6-pyrrolidin-1-ylpyridine-2-carbaldehyde | |
10821162 | |
97% |
97% | |
MFCD08435859 | |
6-pyrrolidin-1-yl picolinaldehyde, 6-pyrrolidin-1-yl pyridine-2-carbaldehyde, 6-pyrrolidin-1-yl-picolinaldehyde, 6-pyrrolidinylpyridine-2-carbaldehyde, 6-1-pyrrolidinyl-2-pyridinecarboxaldehyde, 2-pyridinecarboxaldehyde,6-1-pyrrolidinyl, 2-pyridinecarboxaldehyde, 6-1-pyrrolidinyl | |
O=CC1=NC(=CC=C1)N1CCCC1 | |
176.22 | |
176.22 | |
6-Pyrrolidin-1-ylpyridine-2-carbaldehyde |