Please login to your online account to display your discounted pricing

6-Pyrrolidin-1-ylpyridine-2-carbaldehyde, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC38004CB

 View more versions of this product

Catalog No. CC38004CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.22
Quantity 250mg
Assay 97%
Molecular Formula C10H12N2O
Chemical Name or Material 6-Pyrrolidin-1-ylpyridine-2-carbaldehyde
Assay Percent Range 97%
CAS 230618-24-3

Specifications

Chemical Name or Material 6-Pyrrolidin-1-ylpyridine-2-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H12N2O
Formula Weight 176.22