missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (6-Phthalimidohexyl)triphenylphosphonium bromide, 94%

$123.11 - $123.11

Chemical Identifiers

CAS 129789-59-9
Molecular Formula C32H31BrNO2P
Formula Weight 572.49
MDL Number MFCD04039776
Chemical Name or Material (6-Phthalimidohexyl)triphenylphosphonium bromide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2503406 SDS Alfa Aesar™
B2503406
5g Each for $123.11
AAB2503403 SDS Alfa Aesar™
B2503403
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

129789-59-9
572.49
(6-Phthalimidohexyl)triphenylphosphonium bromide
C32H31BrNO2P
MFCD04039776

Specifications

74° to 76°C
129789-59-9
572.49
(6-Phthalimidohexyl)triphenylphosphonium bromide
5g
C32H31BrNO2P
MFCD04039776
94%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning