missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Phenylnaphthalene-2-boronic acid, 98%

$30.22 - $30.22

Chemical Identifiers

CAS 876442-90-9
Molecular Formula C16H13BO2
Molecular Weight (g/mol) 248.088
MDL Number MFCD26406213
InChI Key XDKCHGWZDWHBQO-UHFFFAOYSA-N
Synonym 6-phenylnaphthalen-2-yl boronic acid, boronic acid, 6-phenyl-2-naphthalenyl, 6-phenylnaphthalene-2-boronic acid, 6-phenylphthalen-2-yl-2-boronic acid, 2-phenylnaphthalen-6-yl boronic acid, 6-phenyl-2-naphthalenyl boronic acid, b-6-phenyl-2-naphthalenyl boronic acid, boronic acid, 6-phenyl-2-naphthalenyl
PubChem CID 57539613
IUPAC Name (6-phenylnaphthalen-2-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)C=C(C=C2)C3=CC=CC=C3)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64331MD View Documents Alfa Aesar™
H64331MD
250mg Each for $30.22
 
Specifications

Chemical Identifiers

876442-90-9
248.088
XDKCHGWZDWHBQO-UHFFFAOYSA-N
57539613
B(C1=CC2=C(C=C1)C=C(C=C2)C3=CC=CC=C3)(O)O
C16H13BO2
MFCD26406213
6-phenylnaphthalen-2-yl boronic acid, boronic acid, 6-phenyl-2-naphthalenyl, 6-phenylnaphthalene-2-boronic acid, 6-phenylphthalen-2-yl-2-boronic acid, 2-phenylnaphthalen-6-yl boronic acid, 6-phenyl-2-naphthalenyl boronic acid, b-6-phenyl-2-naphthalenyl boronic acid, boronic acid, 6-phenyl-2-naphthalenyl
(6-phenylnaphthalen-2-yl)boronic acid

Specifications

250mg
C16H13BO2
11441104
XDKCHGWZDWHBQO-UHFFFAOYSA-N
(6-phenylnaphthalen-2-yl)boronic acid
57539613
Solid
6-Phenylnaphthalene-2-boronic acid
876442-90-9
MFCD26406213
6-phenylnaphthalen-2-yl boronic acid, boronic acid, 6-phenyl-2-naphthalenyl, 6-phenylnaphthalene-2-boronic acid, 6-phenylphthalen-2-yl-2-boronic acid, 2-phenylnaphthalen-6-yl boronic acid, 6-phenyl-2-naphthalenyl boronic acid, b-6-phenyl-2-naphthalenyl boronic acid, boronic acid, 6-phenyl-2-naphthalenyl
B(C1=CC2=C(C=C1)C=C(C=C2)C3=CC=CC=C3)(O)O
248.088
248.09
98%