missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Phenylisocytosine, TCI America™

Manufacturer:  TCI America P21231G

 View more versions of this product

Catalog No. P21231G



Specifications

Specifications

6-Phenylisocytosine
295°C
56741-94-7
MFCD00466412
KXYGHDWFGFZJPJ-UHFFFAOYSA-N
2-amino-6-phenyl-1H-pyrimidin-4-one
95891
Crystal-Powder at 20°C
White-Yellow
1g
C10H9N3O
2-Amino-6-phenyl-4(1H)-pyrimidinone
C1=CC=C(C=C1)C2=CC(=O)N=C(N2)N
187.202
187.20
≥98.0% (HPLC)

Chemical Identifiers

56741-94-7
187.202
KXYGHDWFGFZJPJ-UHFFFAOYSA-N
95891
C1=CC=C(C=C1)C2=CC(=O)N=C(N2)N
C10H9N3O
MFCD00466412
2-Amino-6-phenyl-4(1H)-pyrimidinone
2-amino-6-phenyl-1H-pyrimidin-4-one
Safety and Handling

Safety and Handling

TSCA : No

Documents