Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Phenylhexanoic acid, 98+%

$61.64 - $920.64

Chemical Identifiers

Chemical Name or Material 6-Phenylhexanoic acid
CAS 5581-75-9
Molecular Formula C6H5(CH2)5CO2H
Formula Weight 192.26
MDL Number MFCD00014381
Beilstein 1950106
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1118703

  SDS
alfa aesar™
A1118703
1g Each for $61.64

AAA1118706

  SDS
alfa aesar™
A1118706
5g Each for $215.47

AAA1118714

  SDS
alfa aesar™
A1118714
25g Each for $920.64
Description & Specifications

Specifications

Formula Weight 192.26
Quantity 1g
Assay ≥98%
Beilstein 1950106
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Boiling Point 157° to 160°C (0.3mmHg)
Chemical Name or Material 6-Phenylhexanoic acid
CAS 5581-75-9
Melting Point 17° to 19°C
Molecular Formula C6H5(CH2)5CO2H
Health Hazard 1 Warning
MDL Number MFCD00014381
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

Applications
6-Phenylhexanoic acid was employed as a model compound to investigate the effects of chromophore orientation and molecular conformation on surface-enhanced Raman scattering based on metal nanostructures. It was also employed as a substrate to study the stoichiometry of side chain metabolism or complete mineralization of surrogate napthenic acids under methanogenic conditions

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.