Please login to your online account to display your discounted pricing

6-Oxo-1,6-dihydropyridazine-3-carboxylic acid monohydrate, 97%, Maybridge


Quantity 25g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB07063EB

 View more versions of this product

Catalog No. BTB07063EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 158.12
Quantity 25g
Assay 97%
Chemical Name or Material 6-Hydroxy-3-pyridazinecarboxylic acid monohydrate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H4N2O3·H2O
CAS 37972-69-3
Health Hazard 1 Warning

Specifications

Chemical Name or Material 6-Hydroxy-3-pyridazinecarboxylic acid monohydrate
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H4N2O3·H2O
Formula Weight 158.12
Accessories

Warning