Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid, 97%

$319.43 - $319.43

Chemical Identifiers

Chemical Name or Material 6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
CAS 27372-38-9
Molecular Formula C5H6N2O3
Formula Weight 142.12
MDL Number MFCD00052030
Synonym 1,4,5,6-Tetrahydro-6-oxopyridazine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2028906

  SDS
alfa aesar™
B2028906
5g Each for $319.43
Description & Specifications

Specifications

Formula Weight 142.12
Synonym 1,4,5,6-Tetrahydro-6-oxopyridazine-3-carboxylic acid
Molecular Formula C5H6N2O3
Health Hazard 1 Warning
Assay 97%
CAS 27372-38-9
Melting Point 196° to 198°C
Quantity 5g
Chemical Name or Material 6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00052030
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.