missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-O-tert-Butyldiphenylsilyl-D-glucal, 97%

$44.72 - $141.11

Chemical Identifiers

CAS 87316-22-1
Molecular Formula C22H28O4Si
MDL Number MFCD01321278
Synonym 6-o-tert-butyldiphenylsilyl-d-glucal, 6-o-tert-butyldiphenylsilyl-d-glucose, 6-o-tert-butyl diphenyl silyl-d-glucose, 6-o-tert-butyldiphenylsilyl-d-glucal, 2r,3s,4r,5r-6-tert-butyldiphenylsilyl oxy-2,3,4,5-tetrahydroxyhexanal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60191MD View Documents Alfa Aesar™
H60191MD
250mg Each for $44.72
 
AAH6019103 View Documents Alfa Aesar™
H6019103
1g Each for $141.11
 
Specifications

Chemical Identifiers

87316-22-1
C22H28O4Si
MFCD01321278
6-o-tert-butyldiphenylsilyl-d-glucal, 6-o-tert-butyldiphenylsilyl-d-glucose, 6-o-tert-butyl diphenyl silyl-d-glucose, 6-o-tert-butyldiphenylsilyl-d-glucal, 2r,3s,4r,5r-6-tert-butyldiphenylsilyl oxy-2,3,4,5-tetrahydroxyhexanal

Specifications

+9° (c=1.1 in Chloroform)
250mg
C22H28O4Si
6-o-tert-butyldiphenylsilyl-d-glucal, 6-o-tert-butyldiphenylsilyl-d-glucose, 6-o-tert-butyl diphenyl silyl-d-glucose, 6-o-tert-butyldiphenylsilyl-d-glucal, 2r,3s,4r,5r-6-tert-butyldiphenylsilyl oxy-2,3,4,5-tetrahydroxyhexanal
97%
-9°C
87316-22-1
MFCD01321278
384.54
6-O-tert-Butyldiphenylsilyl-D-glucal
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C