missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-O-tert-Butyldiphenylsilyl-D-glucal, 97%

Manufacturer:  Alfa Aesar™ H60191MD

 View more versions of this product

Catalog No. AAH60191MD



Specifications

Specifications

6-O-tert-Butyldiphenylsilyl-D-glucal
-9°C
87316-22-1
MFCD01321278
384.54
+9° (c=1.1 in Chloroform)
250mg
C22H28O4Si
6-o-tert-butyldiphenylsilyl-d-glucal, 6-o-tert-butyldiphenylsilyl-d-glucose, 6-o-tert-butyl diphenyl silyl-d-glucose, 6-o-tert-butyldiphenylsilyl-d-glucal, 2r,3s,4r,5r-6-tert-butyldiphenylsilyl oxy-2,3,4,5-tetrahydroxyhexanal
97%

Chemical Identifiers

87316-22-1
C22H28O4Si
MFCD01321278
6-o-tert-butyldiphenylsilyl-d-glucal, 6-o-tert-butyldiphenylsilyl-d-glucose, 6-o-tert-butyl diphenyl silyl-d-glucose, 6-o-tert-butyldiphenylsilyl-d-glucal, 2r,3s,4r,5r-6-tert-butyldiphenylsilyl oxy-2,3,4,5-tetrahydroxyhexanal
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

SDS Documents