missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-O-Benzoyl-D-glucal, 97%

$44.72 - $141.11

Chemical Identifiers

CAS 58871-05-9
Molecular Formula C13H14O5
Molecular Weight (g/mol) 250.25
MDL Number MFCD01321275
InChI Key DIEYDTYJOXFFJG-UTUOFQBUSA-N
Synonym 6-o-benzoyl-d-glucal, 2r,3s,4r-3,4-dihydroxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate, 2r,3s,4r-3,4-dihydroxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate, 6-o-benzoylglucal, d-arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, 6-benzoate
PubChem CID 14308105
IUPAC Name [(2R,3S,4R)-3,4-dihydroxy-3,4-dihydro-2H-pyran-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C=CO2)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60465MD View Documents Alfa Aesar™
H60465MD
250mg Each for $44.72
 
AAH6046503 View Documents Alfa Aesar™
H6046503
1g Each for $141.11
 
Specifications

Chemical Identifiers

58871-05-9
250.25
DIEYDTYJOXFFJG-UTUOFQBUSA-N
14308105
C1=CC=C(C=C1)C(=O)OCC2C(C(C=CO2)O)O
C13H14O5
MFCD01321275
6-o-benzoyl-d-glucal, 2r,3s,4r-3,4-dihydroxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate, 2r,3s,4r-3,4-dihydroxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate, 6-o-benzoylglucal, d-arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, 6-benzoate
[(2R,3S,4R)-3,4-dihydroxy-3,4-dihydro-2H-pyran-2-yl]methyl benzoate

Specifications

+44° (c=1.4 in Chloroform)
250mg
C13H14O5
6-o-benzoyl-d-glucal, 2r,3s,4r-3,4-dihydroxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate, 2r,3s,4r-3,4-dihydroxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate, 6-o-benzoylglucal, d-arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, 6-benzoate
C1=CC=C(C=C1)C(=O)OCC2C(C(C=CO2)O)O
250.25
250.25
6-O-Benzoyl-D-glucal
106°C to 111°C
58871-05-9
MFCD01321275
DIEYDTYJOXFFJG-UTUOFQBUSA-N
[(2R,3S,4R)-3,4-dihydroxy-3,4-dihydro-2H-pyran-2-yl]methyl benzoate
14308105
97%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C