missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Nitroquinoline, TCI America™

$55.00 - $210.84

Chemical Identifiers

CAS 613-50-3
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00006799
InChI Key SMHPLBXIVNQFBA-UHFFFAOYSA-N
Synonym quinoline, 6-nitro, 6-nitro-quinoline, ccris 456, 6-nitro quinoline, zlchem 253, pubchem20822, acmc-1bcwl, dsstox_cid_984, 6-nitroquinoline, dsstox_rid_75906
PubChem CID 11945
IUPAC Name 6-nitroquinoline
SMILES C1=CC2=C(C=CC(=C2)[N+](=O)[O-])N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N02525G View Documents TCI America
N02525G
5g Each for $55.00
 
N025225G View Documents TCI America
N025225G
25g Each for $210.84
 
Specifications

Chemical Identifiers

613-50-3
174.159
SMHPLBXIVNQFBA-UHFFFAOYSA-N
11945
C1=CC2=C(C=CC(=C2)[N+](=O)[O-])N=C1
C9H6N2O2
MFCD00006799
quinoline, 6-nitro, 6-nitro-quinoline, ccris 456, 6-nitro quinoline, zlchem 253, pubchem20822, acmc-1bcwl, dsstox_cid_984, 6-nitroquinoline, dsstox_rid_75906
6-nitroquinoline

Specifications

Undesignated
5g
C9H6N2O2
quinoline, 6-nitro, 6-nitro-quinoline, ccris 456, 6-nitro quinoline, zlchem 253, pubchem20822, acmc-1bcwl, dsstox_cid_984, 6-nitroquinoline, dsstox_rid_75906
C1=CC2=C(C=CC(=C2)[N+](=O)[O-])N=C1
174.159
174.16
≥98.0% (GC,T)
151°C
613-50-3
MFCD00006799
SMHPLBXIVNQFBA-UHFFFAOYSA-N
6-nitroquinoline
11945
Crystal-Powder at 20°C
6-Nitroquinoline
Safety and Handling

Safety and Handling

RTECSNumber : VC1900000

TSCA : No