Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Nitroquinoline, 98+%

$83.93 - $311.91

Chemical Identifiers

Chemical Name or Material 6-Nitroquinoline
CAS 613-50-3
Molecular Formula C9H6N2O2
Formula Weight 174.16
MDL Number MFCD00006799
Beilstein 136138
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1721209

  SDS
alfa aesar™
A1721209
10g Each for $83.93

AAA1721218

  SDS
alfa aesar™
A1721218
50g Each for $311.91
Description & Specifications

Specifications

Formula Weight 174.16
Chemical Name or Material 6-Nitroquinoline
Melting Point 151° to 153°C (subl.)
Assay ≥98%
Quantity 10g
Molecular Formula C9H6N2O2
MDL Number MFCD00006799
CAS 613-50-3
Beilstein 136138
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VC1900000
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.